Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF224 Monoclonal Antibody (2C12)
GREENER_CHOICE

Catalog No. PIMA520297
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520297 Supplier Invitrogen™ Supplier No. MA520297
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: HQEWSFQQIW EKIASDLTRS QDLVINSSQF SKEGDFPCQT EAGLSVIHTR QKSSQGNGYK PSFSDVSHFD FHQQLHSGEK SHTCDECGKN FCYISA.

Matrix-assisted laser desorption ionization-time of flight analysis and examination of protein profiles from the SwissProt database revealed that the previously defined p97 repressor is ZNF224, a zinc finger protein. ZNF224, a Kruppel-like zinc finger transcription factor, is the repressor protein that specifically binds to the negative cis-element AldA-NRE and affects the AldA-NRE-mediated transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF224
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C12
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF224
Gene Accession No. Q9NZL3
Gene Alias BMZF2; BMZF-2; Bone marrow zinc finger 2; KOX22; Zinc finger protein 224; zinc finger protein 233; Zinc finger protein 255; zinc finger protein 27; Zinc finger protein KOX22; ZNF224; ZNF233; ZNF255; ZNF27
Gene Symbols ZNF224
Host Species Mouse
Immunogen ZNF224 (NP_037530.1, 95 a.a. ~ 190 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7767
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.