Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF224 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116266
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116266 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116266 Supplier Invitrogen™ Supplier No. PA5116266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF224 Polyclonal Antibody for Western Blot

Matrix-assisted laser desorption ionization-time of flight analysis and examination of protein profiles from the SwissProt database revealed that the previously defined p97 repressor is ZNF224, a zinc finger protein. ZNF224, a Kruppel-like zinc finger transcription factor, is the repressor protein that specifically binds to the negative cis-element AldA-NRE and affects the AldA-NRE-mediated transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF224
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF224
Gene Accession No. Q9NZL3
Gene Alias BMZF2; BMZF-2; Bone marrow zinc finger 2; KOX22; Zinc finger protein 224; zinc finger protein 233; Zinc finger protein 255; zinc finger protein 27; Zinc finger protein KOX22; ZNF224; ZNF233; ZNF255; ZNF27
Gene Symbols ZNF224
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 127-155 amino acids from the N-terminal region of human ZNF224.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.