Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF224 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541138 Supplier Thermo Scientific Supplier No. PA541138
Add to Cart
Add to Cart

ZNF224 Polyclonal Antibody

ZNF224 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF224
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF224
Gene Accession No. Q9NZL3
Gene Alias BMZF2; BMZF-2; Bone marrow zinc finger 2; KOX22; Zinc finger protein 224; zinc finger protein 233; Zinc finger protein 255; zinc finger protein 27; Zinc finger protein KOX22; ZNF224; ZNF233; ZNF255; ZNF27
Gene Symbols ZNF224
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF224 (aa 124-173).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7767
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.