Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF227 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF227 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ZNF227 Polyclonal specifically detects ZNF227 in Human samples. It is validated for Western Blot.Specifications
ZNF227 | |
Polyclonal | |
Purified | |
RUO | |
AAO45835 | |
7770 | |
Synthetic peptide directed towards the middle region of human ZNF227. Peptide sequence ESGDCFNKSSFHSYQSNHTGEKSYRCDSCGKGFSSSTGLIIHYRTHTGEK. | |
Primary | |
30 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
KIAA1084, zinc finger protein 507 | |
ZNF227 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title