Learn More
Invitrogen™ ZNF23 Monoclonal Antibody (2D3)

Description
Peptide Sequence: GLQGLQTDIQ TDNDLTKEMY EGKENVSFEL QRDFSQETDF SEASLLEKQQ EVHSAGNIKK EKSNTIDGTV KDETSPVEEC FFSQSSNSYQ CHTITGEQP.
Specifications
Specifications
| Antigen | ZNF23 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2D3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF23 |
| Gene Accession No. | P17027 |
| Gene Alias | B230354B21Rik; KOX16; kruppel-like zinc finger factor X31; Zfp23; Zfp612; zinc finger protein 23; zinc finger protein 23 (KOX 16); zinc finger protein 32; zinc finger protein 359; Zinc finger protein 612; Zinc finger protein KOX16; ZNF23; ZNF359; ZNF612 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.