Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103264 Supplier Invitrogen™ Supplier No. PA5103264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNF23.

ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF23
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNF23
Gene Accession No. P17027
Gene Alias B230354B21Rik; KOX16; kruppel-like zinc finger factor X31; Zfp23; Zfp612; zinc finger protein 23; zinc finger protein 23 (KOX 16); zinc finger protein 32; zinc finger protein 359; Zinc finger protein 612; Zinc finger protein KOX16; ZNF23; ZNF359; ZNF612
Gene Symbols ZNF23
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNF23(Accession P17027), corresponding to amino acid residues R166-R216.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7571
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.