Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF230 Antibody (OTI1E2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274934FR
Description
ZNF230 Monoclonal specifically detects ZNF230 in Human samples. It is validated for Immunohistochemistry.Specifications
ZNF230 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ZNF230 | |
Full length recombinant protein of human ZNF230 (NP_006291) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI1E2 | |
Immunohistochemistry | |
FLJ12515, FLJ14855, zinc finger protein 502 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
7773.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction