Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF230 Monoclonal Antibody (OTI1E2)
GREENER_CHOICE

Catalog No. PIMA527089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527089 Supplier Invitrogen™ Supplier No. MA527089
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF230 Monoclonal Antibody for IHC (P)

ZNF230 is predicted to enable DNA-binding transcription factor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. This gene is predicted to be active in the nucleus and is predicted to be involved in regulation of transcription by RNA polymerase II. ZN230 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF230
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF230
Gene Accession No. Q9UIE0
Gene Alias 2610110L04Rik; AA792898; AU022812; C3HC4-like zinc finger protein; FDZF2; MGC8715; RING finger protein 141; Rnf141; RNF141 protein; ZFP26; ZFP36; Zinc finger protein 230; Zinc Finger Protein FDZF2; ZNF230
Gene Symbols ZNF230
Host Species Mouse
Immunogen Full length human recombinant protein of ZNF230 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7773
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.