Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF230 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564906
Description
Immunogen sequence: GKTIAEAGPHE DCPCQQIWEQ TASDLTQSQD SIINNSHFFE QGDVPSQVEA GLSIIHTGQK PSQNGKCKQS FSDVAIFDPP QQFHSGEKS Highest antigen sequence identity to the following orthologs - mouse 29%, rat 29%.
ZNF230 is predicted to enable DNA-binding transcription factor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity. This gene is predicted to be active in the nucleus and is predicted to be involved in regulation of transcription by RNA polymerase II. ZN230 may be involved in transcriptional regulation.
Specifications
ZNF230 | |
Polyclonal | |
Unconjugated | |
ZNF230 | |
2610110L04Rik; AA792898; AU022812; C3HC4-like zinc finger protein; FDZF2; MGC8715; RING finger protein 141; Rnf141; RNF141 protein; ZFP26; ZFP36; Zinc finger protein 230; Zinc Finger Protein FDZF2; ZNF230 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIE0 | |
ZNF230 | |
Recombinant Human ZNF230. Recombinant protein control fragment (Product #RP-104551). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction