Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF141 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582615
Description
Immunogen sequence: QLVINKLPEK VAKHVTLVRE SGSLTYEEFL GRVAELNDVT AKVASGQEKH LLFEVQPGSD SSAFWKVVVR VVCTKINKSS GIVEASRIMN LYQFIQLYKD ITSQAAGVLA QSSTSEEPDE NSSSVTSCQA SLWMGRVKQL TDE.
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. Abundant expression of this gene was found in the testicular tissue of fertile men, but was not detected in azoospermic patients. Studies of the mouse counterpart suggest that this gene may function as a testis specific transcription factor during spermatogenesis.
Specifications
RNF141 | |
Polyclonal | |
Unconjugated | |
RNF141 | |
RFP141; Ring Finger Protein 141; ZFP26; ZNF230 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVD5 | |
RNF141 | |
Recombinant protein corresponding to Human RNF141. Recombinant protein control fragment (Product #RP-91617). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction