Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF232 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555578 Supplier Invitrogen™ Supplier No. PA555578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SWYEPSAELV QTRMAVSLTA AETLALQGTQ GQEKMMMMGP KEEEQSCEYE TRLPGNHSTS QEIFRQRFRH LRYQETPGPR EALSQLRVLC CEWLRPEKHT KEQILEFL Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 40%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF232, also known as Zinc finger and SCAN domain-containing protein 11, is a 417 amino acid protein belonging to the Kruppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF232 contains one SCAN box domain and five C2H2-type zinc fingers. Due to the presence of these domains, ZNF232 may be involved in transcriptional regulation. Ubiquitously expressed, ZNF232 is present at high levels in testis, liver and ovary. ZNF232 exists as two isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF232
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF232
Gene Accession No. Q9UNY5
Gene Alias Zinc finger and SCAN domain-containing protein 11; zinc finger protein 232; ZNF232; ZSCAN11
Gene Symbols ZNF232
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF232. Recombinant protein control fragment (Product #RP-89446).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7775
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.