Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF234 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118372
This item is not returnable.
View return policy
Description
ZNF234 Polyclonal Antibody for Western Blot, IHC (P), IP
May be involved in transcriptional regulation.
Specifications
ZNF234 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q14588 | |
ZNF234 | |
E. coli-derived Human ZNF234 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
ZNF234 | |
C2-H2 type zinc finger protein; Homo sapiens zinc finger 234; HZF4; zinc finger protein 234; zinc finger protein 234, partial sequence; zinc finger protein 269; zinc finger protein 4; zinc finger protein HZF4; ZNF234; ZNF269 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10780 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction