Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF237 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554273 Supplier Invitrogen™ Supplier No. PA554273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEKTSQLQLS VECGTDTLLI QENVNLPPSS TSTIADTFQE QLEEKNFEDS IVPVVLSADP GTWPRILNIK QRDTLVENVP Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 51%.

ZNF237 is a candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF237
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYM5
Gene Accession No. Q9UJ78
Gene Alias HSPC050; MYM; zinc finger MYM-type containing 5; zinc finger MYM-type protein 5; Zinc finger protein 198-like 1; zinc finger protein 237; zinc finger, MYM-type 5; ZMYM5; ZNF198L1; ZNF237
Gene Symbols ZMYM5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF237. Recombinant protein control fragment (Product #RP-91629).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.