Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF238 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100372 Supplier Thermo Scientific Supplier No. PA5100372
Add to Cart
Add to Cart

ZNF238 Polyclonal Antibody

ZNF238 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF238
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZBTB18
Gene Accession No. Q99592, Q9JKY3, Q9WUK6
Gene Alias 58 kDa repressor protein; BTB/POZ domain protein ZF238; C2H2-171; MRD22; novel zinc finger domain containing protein; RP58; rRP58; si:ch211-221n23.1; TAZ1; TAZ-1; Transcriptional repressor RP58; transcriptional represssor; translin-associated zinc finger protein 1; Zbtb18; Zfp238; Zfp-238; zinc finger and BTB domain containing 18; zinc finger and BTB domain-containing protein 18; Zinc finger protein 238; zinc finger protein C2H2-171; Znf238
Gene Symbols ZBTB18
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZBTB18(Accession Q99592), corresponding to amino acid residues T228-A278.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10472, 30928, 64619
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.