Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF24 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547521
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintenance in the progenitor stage by promoting the cell cycle. Specifically binds to the 5'-TCAT-3' DNA sequence. Has transcription repressor activity in vitro.
Specifications
ZNF24 | |
Polyclonal | |
Unconjugated | |
Znf24 | |
3526401F17Rik; 5033419P20Rik; AI480505; AI642085; AL024422; Hmcns; Hypomyelinated CNS protein; KOX17; LOW QUALITY PROTEIN: zinc finger protein 24; retinoic acid suppression protein A; RSG-A; ZF-12; Zfp191; zfp-191; Zfp24; Zinc finger and SCAN domain-containing protein 3; zinc finger protein 191; Zinc finger protein 24; zinc finger protein 24 (KOX 17); zinc finger protein 24-like protein; zinc finger protein KOX17; Zinc finger protein ZF-12; zinc finger ZF-12; Znf191; Znf24; Znp191; ZSCAN3 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
7572 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P17028 | |
Znf24 | |
E. coli-derived recombinant human ZNF24 Ser2-Val368. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction