Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726978 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6978 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6978 Supplier Proteintech Group Inc Supplier No. 112191AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintece in the progenitor stage by promoting the cell cycle. Specifically binds to the 5′-TCAT-3′ DNA sequence. Has transcription repressor activity in vitro.

Specifications

Antigen ZNF24
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF24
Gene Accession No. P17028, Q7TNK3, Q91VN1
Gene Alias KOX17, RSG A, Zfp191, Zinc finger protein 191, zinc finger protein 24, Zinc finger protein KOX17, ZNF191, ZNF24, ZSCAN3
Gene Symbols Znf24
Host Species Rabbit
Immunogen ZNF24 Fusion Protein Ag1724
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360204, 59057, 7572
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.