Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF248 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553804 Supplier Invitrogen™ Supplier No. PA553804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEYGEIFCDN SAFIIHQGAY TRKILREYKV SDKTWEKSAL LKHQIVHMGG KSYDYNENGS NFSKKS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 67%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF248
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF248
Gene Accession No. Q8NDW4
Gene Alias bA162G10.3; bA162G10.3 (zinc finger protein); KRAB protein domain; Kruppel-type zinc finger protein; zinc finger protein 248; ZNF248
Gene Symbols ZNF248
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF248. Recombinant protein control fragment (Product #RP-91704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57209
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.