Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF253 Monoclonal Antibody (1D7)
GREENER_CHOICE

Catalog No. PIMA522272
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522272 Supplier Invitrogen™ Supplier No. MA522272
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: QNSFQIGMLR RYEECRHDNL QLKKGCKSVG EHKVHKGGYN GLNQCLTTTQ KEIFQCDKYG KVFHKFSNSN TYKTRHTGIN LFKCIICGKA.

ZNF253 may function as a transcription factor. Seems to have a transcriptional repression activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF253
Applications ELISA
Classification Monoclonal
Clone 1D7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF253
Gene Accession No. O75346
Gene Alias BMZF1; BMZF-1; Bone marrow zinc finger 1; DNA-binding protein; zinc finger protein 253; Zinc finger protein 411; ZNF253; ZNF411
Gene Symbols ZNF253
Host Species Mouse
Immunogen ZNF253 (NP_066385.2, 91 a.a. ~ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56242
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.