Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF253 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541615 Supplier Invitrogen™ Supplier No. PA541615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LTTHKRIHTG EKPYKCEECG KAFNWSSDLN KHKKIHIERK PYIVKNVTDL Sequence homology: Cow: 83%; Horse: 83%; Human: 100%; Zebrafish: 83%.

ZNF253 may function as a transcription factor. Seems to have a transcriptional repression activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF253
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF253
Gene Accession No. O75346
Gene Alias BMZF1; BMZF-1; Bone marrow zinc finger 1; DNA-binding protein; zinc finger protein 253; Zinc finger protein 411; ZNF253; ZNF411
Gene Symbols ZNF253
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF253 (aa 439-488).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56242
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.