Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF256 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563118 Supplier Invitrogen™ Supplier No. PA563118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLTLTTSLGG SGAGDEEAPY QQSTSPQRVS QVRIPKALPS PQKTNPCEIC Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 39%.

Transcriptional repressor that plays a role in cell proliferation. Requires TRIM28 for its activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF256
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF256
Gene Accession No. Q9Y2P7
Gene Alias BMZF3; BMZF-3; Bone marrow zinc finger 3; zinc finger protein 256; ZNF256
Gene Symbols ZNF256
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF256. Recombinant protein control fragment (Product #RP-101370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.