Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF256 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501726243 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6243 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6243 Supplier Proteintech Group Inc Supplier No. 103601AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional repressor that plays a role in cell proliferation. Requires TRIM28 for its activity.

Specifications

Antigen ZNF256
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF256
Gene Accession No. Q9Y2P7
Gene Alias BMZF 3, BMZF3, Bone marrow zinc finger 3, zinc finger protein 256, ZNF256
Gene Symbols ZNF256
Host Species Rabbit
Immunogen ZNF256 Fusion Protein Ag0390
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10172
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.