Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF258 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567211 Supplier Invitrogen™ Supplier No. PA567211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGFIICPGSKE SSPRPQCVIC GEILSSENMK PANLSHHLKT KHSELENKPV DFFEQKSLEM ECQNSSLKKC LLVEKSLVKA SYLIAFQTAA S Highest antigen sequence identity to the following orthologs - mouse 80%, rat 82%.

ZNF258 is part of a novel, putative, zinc-binding motif (MYM) family which encodes proteins that maintain the repeats of the MYM motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF258
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZMYM6
Gene Accession No. O95789
Gene Alias Buster2; KIAA1353; MYM; transposase-like protein; transposon-derived Buster2 transposase-like protein; ZBED7; zinc finger MYM-type containing 6; zinc finger MYM-type protein 6; Zinc finger protein 258; zinc finger, BED-type containing 7; zinc finger, MYM-type 6; zinc finger, MYM-type containing 6; ZMYM6; ZNF198L4; ZNF258
Gene Symbols ZMYM6
Host Species Rabbit
Immunogen Recombinant Human ZNF258. Recombinant protein control fragment (Product #RP-107845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.