Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF259 Antibody (SR1907), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322465100UL
Description
ZNF259 Monoclonal antibody specifically detects ZNF259 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ZNF259 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
8882 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
SR1907 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
ERP1, EZFIT-related protein 1, KIAA1396MGC150657, MGC150658, Z1971, zinc finger protein 471 | |
A synthesized peptide derived from human ZNF259 (Uniprot #: O75312) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction