Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF259 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501730316 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0316 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0316 Supplier Proteintech Group Inc Supplier No. 155581AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The MW of this protein is 51 kDa, and this antibody specifically recognizes the 51 kDa protein.

ZNF259, also named as Zpr1, is an evolutionarily conserved protein characterized by two C4 zinc fingers and two Zpr1-specific conserved homology domains. Zpr1 was first identified in mammalian cells as a cytoplasmic protein that was capable of binding the intracellular domain of the inactivated EGF receptor (EGFR).

Specifications

Antigen ZNF259
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZPR1
Gene Accession No. O75312
Gene Alias zinc finger protein 259, Zinc finger protein ZPR1, ZNF259, ZPR1
Gene Symbols ZPR1
Host Species Rabbit
Immunogen ZNF259 Fusion Protein Ag7980
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8882
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.