Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF263 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557149 Supplier Invitrogen™ Supplier No. PA557149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPPEGNMEDK EMTGPQLPES LEDVAMYISQ EEWGHQDPSK RALSRDTVQE SYENVDSLES HIPSQEVPGT QVGQGGKLWD PSVQSCKEG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

Might play an important role in basic cellular processes as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF263
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF263
Gene Accession No. O14978
Gene Alias FPM315; zinc finger protein 263; Zinc finger protein FPM315; zinc finger protein with KRAB and SCAN domains 12; ZKSCAN12; ZNF263; ZSCAN44
Gene Symbols ZNF263
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF263. Recombinant protein control fragment (Product #RP-97587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.