Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF263 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982043
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2043 Supplier Bioss Supplier No. BS12217R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Might play an important role in basic cellular processes as a transcriptional repressor.

Specifications

Antigen ZNF263
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF263
Gene Accession No. O14978
Gene Alias FPM315; Zinc finger protein 263; Zinc finger protein FPM315; Zinc finger protein with KRAB and SCAN domains 12; ZKSCAN12; ZNF263; ZSCAN44
Gene Symbols ZNF263
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF263.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10127
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.