Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF264 Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF810500


Description
ZNF264 Monoclonal antibody specifically detects ZNF264 in Human samples. It is validated for Western BlotSpecifications
ZNF264 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86UA6 | |
Mouse | |
Protein A/G | |
RUO | |
84268 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI8G5 | |
Unconjugated | |
RPAIN | |
KIAA0412; Zinc finger protein 264; ZNF264 | |
Full length human recombinant protein of human RPAIN produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction