Learn More
ZNF264 Mouse anti-Human, Clone: OTI10F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810494

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.Specifications
ZNF264 | |
Monoclonal | |
Unconjugated | |
ZNF264 | |
zinc finger protein 264 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9422 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI10F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43296 | |
ZNF264 | |
Full length human recombit protein of human ZNF264 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.