Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF264 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596651 Supplier Invitrogen™ Supplier No. PA596651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAVLTDRA QVSVTFDDVA VTFTKEEWGQ LDLAQRTLYQ EVMLENCGLL VSLGCPVPKA ELICHLEHGQ EPWTRKEDLS QDTCPGDKGK PKTTEPTTCE PALSEGISLQ GQVTQGNSVD SQLGQAEDQD GLSEMQEGHF RPGIDPQEKS PGKMSPECDG LGTADGVCSR IGQEQVSPGD RVRSHNSCES GKDPMIQEEE NNFKCSECGK VFNKKHLLAG; Positive Samples: Rat serum; Cellular Location: Nucleus.

ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF264
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Alias KIAA0412; Zinc finger protein 264; ZNF264
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZNF264 (NP_0034081).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.