Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF266 Monoclonal Antibody (4G5)
GREENER_CHOICE

Catalog No. PIMA521156
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521156 Supplier Invitrogen™ Supplier No. MA521156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GRAFTVSSCL SQHMKIHVGE KPYECKECGI AFTRSSQLTE HLKTHTAKDP FECKICGKSF RNSSCLSDHF RIH.

This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF266
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF266
Gene Accession No. Q14584
Gene Alias HZF1; KIAA2007; zinc finger protein 1; zinc finger protein 266; Zinc finger protein HZF1; ZNF266
Gene Symbols ZNF266
Host Species Mouse
Immunogen ZNF266 (NP_006622.2, 274 a.a. ~ 346 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10781
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.