Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF266 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562921 Supplier Invitrogen™ Supplier No. PA562921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSENTFECYL YGVDFLTLHK KTSTGEQRSV FSQC Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 44%.

This gene encodes a protein containing many tandem zinc-finger motifs. Zinc fingers are protein or nucleic acid-binding domains, and may be involved in a variety of functions, including regulation of transcription. This gene is located in a cluster of similar genes encoding zinc finger proteins on chromosome 19. Alternative splicing results in multiple transcript variants for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF266
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF266
Gene Accession No. Q14584
Gene Alias HZF1; KIAA2007; zinc finger protein 1; zinc finger protein 266; Zinc finger protein HZF1; ZNF266
Gene Symbols ZNF266
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF266. Recombinant protein control fragment (Product #RP-101491).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.