Learn More
Invitrogen™ ZNF268 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588851
Description
Immunogen sequence: MATRVRTASI WVPPLQERNS SWDRIRKLQG QESILGQGTP GLQPLPGTPR QKQKSRRIEK VLEWLFISQE QPKITKSWGP LSFMDVFVDF TWEEWQLLDP AQKCLYRSVM LENYSNLVSL GYQHTKPDII FKLEQGEELC MVQAQVPNQT CPNTVWKIDD LMDWHQENKD KLGSTAKSFE CTTFGKLCLL STKYLSRQKP HKCGTHGKSL KYIDFTSDYA RNNPNGFQVH GKSFFHSKHE QTVIGIKYCE SIESGKTVNK KSQLMCQQMY; Positive Samples: Mouse kidney, Rat ovary; Cellular Location: Cytoplasm, Nucleus, Nucleus.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 268 (ZNF268), also known as zinc finger protein 3 or HZF3, is a 947 amino acid protein belonging to the krueppel C2H2-type zinc-finger protein family. ZNF268 contains 24 C2H2-type zinc fingers and one KRAB domain. Localized to the nucleus, ZNF268 is involved in transcriptional regulation and is highly expressed in three to five week old embryos. ZNF268 has been implicated in human leukemia, due to the identification of an alternatively spliced form in leukemia patients. Two named isoforms of ZNF268 exist as a result of alternative splicing events.
Specifications
ZNF268 | |
Polyclonal | |
Unconjugated | |
ZNF268 | |
HZF3; zinc finger protein 268; zinc finger protein 3; zinc finger protein HZF3; ZNF268 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10795 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.28 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q14587 | |
ZNF268 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_0011593531). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.