Learn More
Invitrogen™ ZNF268 Polyclonal Antibody

Description
ZNF268 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | ZNF268 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | ZNF268 |
| Gene Accession No. | Q14587 |
| Gene Alias | HZF3; zinc finger protein 268; zinc finger protein 3; zinc finger protein HZF3; ZNF268 |
| Gene Symbols | ZNF268 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.