Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF268 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980399
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0399 Supplier Bioss Supplier No. BS0625R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 268 (ZNF268), also known as zinc finger protein 3 or HZF3, is a 947 amino acid protein belonging to the krueppel C2H2-type zinc-finger protein family. ZNF268 contains 24 C2H2-type zinc fingers and one KRAB domain. Localized to the nucleus, ZNF268 is involved in transcriptional regulation and is highly expressed in three to five week old embryos. ZNF268 has been implicated in human leukemia, due to the identification of an alternatively spliced form in leukemia patients. Two named isoforms of ZNF268 exist as a result of alternative splicing events.

Specifications

Antigen ZNF268
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF268
Gene Accession No. Q14587
Gene Alias HZF3; Zinc finger protein 268; zinc finger protein 3; Zinc finger protein HZF3; ZNF268
Gene Symbols ZNF268
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF268.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10795
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.