Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF271 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568710 Supplier Invitrogen™ Supplier No. PA568710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 85%; Human: 100%; Mouse: 85%; Rat: 83%.

Zinc finger proteins interact with nucleic acids and have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF271
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF271P
Gene Accession No. 0
Gene Alias CT-ZFP48; HZF7; mszf13; mszf20-1; mszf28; mszf68; Zfp239; Zfp35; Zfp-35; zinc finger protein 239; zinc finger protein 271; zinc finger protein 271, pseudogene; zinc finger protein 35; ZNF271; ZNF271P; ZNF-dp; ZNFEB; ZNF-EB
Gene Symbols ZNF271P
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human ZNF271.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10778
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.