Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF273 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585037 Supplier Invitrogen™ Supplier No. PA585037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLPAGIGRST AKTPGLPGSL E Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 48%.

This gene is a member of the krueppel C2H2-type zinc-finger protein family and encodes a protein with 13 C2H2-type zinc fingers and a KRAB domain. This nuclear protein is involved in transcriptional regulation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF273
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF273
Gene Accession No. Q14593
Gene Alias HZF9; Zinc finger protein 273; zinc finger protein 9; zinc finger protein HZF9; ZNF273
Gene Symbols ZNF273
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF273. Recombinant protein control fragment (Product #RP-108686).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.