Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF273 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569709
Description
This target displays homology in the following species: Human: 100%.
This gene is a member of the krueppel C2H2-type zinc-finger protein family and encodes a protein with 13 C2H2-type zinc fingers and a KRAB domain. This nuclear protein is involved in transcriptional regulation. Alternative splicing results in multiple transcript variants.
Specifications
ZNF273 | |
Polyclonal | |
Unconjugated | |
ZNF273 | |
HZF9; Zinc finger protein 273; zinc finger protein 9; zinc finger protein HZF9; ZNF273 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10793 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14593 | |
ZNF273 | |
synthetic peptide directed towards the C-terminal region of human ZNF273. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction