Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF274 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556001
Description
Immunogen sequence: FGHLVSVGWE TTLENKELAP NSDIPEEEPA PSLKVQESSR DCALSSTLED TLQGGVQEVQ DTVLKQMESA QEKDLPQKKH FDNRESQANS GALDTNQVSL QKIDNPESQA NSGALDTNQV LLHKIPPRKR LRKRDSQVKS MK Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 27%.
ZNF274 is a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus.This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals.
Specifications
ZNF274 | |
Polyclonal | |
Unconjugated | |
ZNF274 | |
DKFZp686K08243; HFB101; KRAB zinc finger protein HFB101; neurotrophin receptor-interacting factor homolog; SP2114; Zf2; zinc finger protein 274; Zinc finger protein HFB101; zinc finger protein with KRAB and SCAN domains 19; Zinc finger protein zfp2; ZKSCAN19; ZNF274; ZSCAN51 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10782 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GC6 | |
ZNF274 | |
Recombinant protein corresponding to Human ZNF274. Recombinant protein control fragment (Product #RP-99755). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction