Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF276 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103342C
Description
ZNF276 Polyclonal antibody specifically detects ZNF276 in Human, Mouse samples. It is validated for Western BlotSpecifications
ZNF276 | |
Polyclonal | |
Western Blot | |
DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF | |
The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human zinc finger protein 276 using the numbering given in entry NP_001106997.1 (GeneID 92822). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
92822 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction