Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF276 Antibody (PCRP-ZNF276-1A5) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314066JF635
Description
ZNF276 Monoclonal antibody specifically detects ZNF276 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZNF276 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF | |
| Recombinant full-length human ZNF276 protein (Uniprot: Q8N554) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-ZNF276-1A5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 92822 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction