Learn More
Invitrogen™ ZNF276 Polyclonal Antibody

Description
ZNF276 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ZNF276 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ZNF276 |
| Gene Accession No. | Q8N554 |
| Gene Alias | CENPZ; CENP-Z; centromere protein Z; ZADT; ZFP276; Zfp-276; Zinc finger protein 276; zinc finger protein 276 homolog; zinc finger protein 477; zinc finger, AD-type; ZNF276; ZNF477 |
| Gene Symbols | ZNF276 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.