Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF276 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554376 Supplier Invitrogen™ Supplier No. PA554376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKDEFSDLSE GDVLSEDEND KKQNAQSSDE SFEPYPERKV SGKKSESKEA KKSEEPRIRK KPGPKPGWKK KLRCEREELP TIY Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF276
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF276
Gene Accession No. Q8N554
Gene Alias CENPZ; CENP-Z; centromere protein Z; ZADT; ZFP276; Zfp-276; Zinc finger protein 276; zinc finger protein 276 homolog; zinc finger protein 477; zinc finger, AD-type; ZNF276; ZNF477
Gene Symbols ZNF276
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF276. Recombinant protein control fragment (Product #RP-93589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.