Learn More
Invitrogen™ ZNF276 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113625
Description
Immunogen sequence: LSEGDVLSED ENDKKQNAQS SDESFEPYPE RKVSGKKSES KEAKKSEEPR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 85%; Rat: 93%.
May be involved in transcriptional regulation.
Specifications
| ZNF276 | |
| Polyclonal | |
| Unconjugated | |
| ZNF276 | |
| CENPZ; CENP-Z; centromere protein Z; ZADT; ZFP276; Zfp-276; Zinc finger protein 276; zinc finger protein 276 homolog; zinc finger protein 477; zinc finger, AD-type; ZNF276; ZNF477 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 92822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N554 | |
| ZNF276 | |
| Synthetic peptide directed towards the middle region of human ZNF276. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.