Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF277 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555577 Supplier Invitrogen™ Supplier No. PA555577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPVAEQDKLL KHMIIEHKIV IADVKLVADF QRYILYWRKR FTEQPITDFC SVIRINSTAP FEEQENYFLL CDVLPEDRIL REELQKQRLR EILEQQQQER NDTNFHGVCM FCNEEFLGNR SVILNH Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 93%.

ZNF277 (Zinc Finger Protein 277) is a Protein Coding gene. Diseases associated with ZNF277 includeSpecific Language ImpairmentandSpeech And Communication Disorders. Gene Ontology (GO) annotations related to this gene includeRNA polymerase II core promoter sequence specific DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF277
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF277
Gene Accession No. Q9NRM2
Gene Alias NRIF4; Nuclear receptor-interacting factor 4; zinc finger protein (C2H2 type) 277; zinc finger protein 277; zinc finger protein 277 pseudogene; ZNF277; ZNF277P
Gene Symbols ZNF277
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF277. Recombinant protein control fragment (Product #RP-89439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.