Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF277 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110599 Supplier Invitrogen™ Supplier No. PA5110599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASKTQGAV ARMQEDRDGS CSTVGGVGYG DSKDCILEPL SLPESPGGTT TLEGSPSVPC IFCEEHFPVA EQDKLLKHMI IEHKIVIADV KLVADFQRYI LYWRKRFTEQ PITDFCSVIR INSTAPFEEQ ENYFLLCDVL PEDRILREEL QKQRLREILE QQQQERNDTN FHGVCMFCNE EFLGNRSVIL NHMAREHAFN IGLPDNIVNC NEFLCTLQKK.

ZNF277 (Zinc Finger Protein 277) is a Protein Coding gene. Diseases associated with ZNF277 includeSpecific Language ImpairmentandSpeech And Communication Disorders. Gene Ontology (GO) annotations related to this gene includeRNA polymerase II core promoter sequence specific DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF277
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF277
Gene Accession No. Q9NRM2
Gene Alias NRIF4; Nuclear receptor-interacting factor 4; zinc finger protein (C2H2 type) 277; zinc finger protein 277; zinc finger protein 277 pseudogene; ZNF277; ZNF277P
Gene Symbols ZNF277
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZNF277 (NP_068834.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11179
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.