Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF280A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560724 Supplier Invitrogen™ Supplier No. PA560724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HFRQYPAHVS QPANHVTSMA KAIMPVSLSE GRSTDSPVTM KSSSEPGYKM SSPQVVSPNY SDSLPPGTQC LVGAMVSGGG RNESSPDSKR LSTSDINSRD SKRVKLRDGI PGVPSLAVVP SDMSSTISTN TPSQGICNSS NHVQNGVTFP WPDANGKA Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 35%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. SUHW1 (Suppressor of hairy wing homolog 1), also known as ZNF280A (Zinc finger protein 280A), 3'OY11.1, ZNF636 or ZNF280, is a 542 amino acid protein that contains four C2H2-type zinc fingers. Localized to the nucleus, SUHW1 is thought to function as a transcription factor that may mediate transcriptional regulation events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF280A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF280A
Gene Accession No. P59817
Gene Alias 3'OY11.1; SUHW1; Suppressor of hairy wing homolog 1; zinc finger protein 280; zinc finger protein 280A; Zinc finger protein 636; ZNF280; ZNF280A; ZNF636
Gene Symbols ZNF280A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF280A. Recombinant protein control fragment (Product #RP-91334).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.