Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF280A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539486
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. SUHW1 (Suppressor of hairy wing homolog 1), also known as ZNF280A (Zinc finger protein 280A), 3′OY11.1, ZNF636 or ZNF280, is a 542 amino acid protein that contains four C2H2-type zinc fingers. Localized to the nucleus, SUHW1 is thought to function as a transcription factor that may mediate transcriptional regulation events.
Specifications
ZNF280A | |
Polyclonal | |
Unconjugated | |
ZNF280A | |
Zinc finger protein 280A, 3′OY11.1, Suppressor of hairy wing homolog 1, Zinc finger protein 636, ZNF280A, SUHW1, ZNF280, ZNF636 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129025 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59817 | |
ZNF280A | |
A synthetic peptide derived from the internal region of human ZNF280A | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction