Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF280B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568339
Description
Predicted to react with Mouse samples.
May function as a transcription factor.
Specifications
ZNF280B | |
Polyclonal | |
Unconjugated | |
ZNF280B | |
5'OY11.1; D87009.C22.3; SUHW2; Suppressor of hairy wing homolog 2; Unknown (protein for MGC:127580); zinc finger protein 279; Zinc finger protein 280B; zinc finger protein 632; ZNF279; ZNF280B; ZNF632 | |
Rabbit | |
affinity chromatography | |
RUO | |
140883 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86YH2 | |
ZNF280B | |
Synthesized peptide derived from human ZNF280B. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction