Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF280B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569037 Supplier Invitrogen™ Supplier No. PA569037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Human: 100%; Pig: 92%.

May function as a transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF280B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF280B
Gene Accession No. Q86YH2
Gene Alias 5'OY11.1; D87009.C22.3; SUHW2; Suppressor of hairy wing homolog 2; Unknown (protein for MGC:127580); zinc finger protein 279; Zinc finger protein 280B; zinc finger protein 632; ZNF279; ZNF280B; ZNF632
Gene Symbols ZNF280B
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human ZNF280B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140883
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.