Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF280B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563742
Description
Immunogen sequence: DSPIIIEPLS KPDYRNSSPQ VVPNNSSELP SPLITFTDSL HHPVSTALSV GGINESPRVS KQLSTFEVNS INPKRAKLRD GII Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 47%.
May function as a transcription factor.
Specifications
ZNF280B | |
Polyclonal | |
Unconjugated | |
ZNF280B | |
5'OY11.1; D87009.C22.3; SUHW2; Suppressor of hairy wing homolog 2; Unknown (protein for MGC:127580); zinc finger protein 279; Zinc finger protein 280B; zinc finger protein 632; ZNF279; ZNF280B; ZNF632 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140883 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YH2 | |
ZNF280B | |
Recombinant protein corresponding to Human ZNF280B. Recombinant protein control fragment (Product #RP-103839). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction